Anti-IgGT Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB112874-1
Supplier: Abcam
Specifications
- Antibody type:Secondary
- Antigen symbol:IgGT
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:1 mg
Specifications
About this item
Type: Secondary
Antigen: Horse IgGT
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: