Anti-IGFBP6 Goat Polyclonal Antibody
Catalog # ABCAAB109765-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Insulin-like Growth Factor Binding Protein 6
- Antigen symbol:IGFBP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: IGFBP6 (insulin-like growth factor binding protein 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human