Anti-Nav1.6/SCN8A Rabbit Polyclonal Antibody
ABCAAB65166-100
: Abcam
- Antibody type:Primary
- Antigen name:Sodium channel, voltage gated, type VIII, alpha subunit
- Antigen symbol:SCN8A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Seq:1286
- Size:100 µl
- Pack type:Vial
- Pk:100 µl
Rabbit polyclonal to Nav1.6/SCN8A.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Nav1.6/SCN8A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat