Anti-DES Mouse monoclonal antibody unconjugated
Catalog # BIRBORB87455-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:desmin
- Antigen symbol:DES
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, 0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- ELISA: 1:10000; Immunochemistry: 1:1000-1:2000; Immunochemistry: 1:1000-1:2000; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: DES (desmin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human