Order Entry
Finland
ContactUsLinkComponent
Anti-SLC34A2 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)
Anti-SLC34A2 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)
Catalog # BIRBORB130629-100
Supplier:  Biorbyt
Anti-SLC34A2 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)
Catalog # BIRBORB130629-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 34 (type II sodium/phosphate contransporter), member 2
  • Antigen symbol:
    Solute carrier family 34, member 2
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)/Cy5.5®
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    77 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: SLC34A2 (solute carrier family 34 (type II sodium/phosphate contransporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat