Anti-Influenza A, Avian, H5N1, Hemagglutinin Rabbit Polyclonal Antibody
BIRBORB109514-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Influenza A, Avian, H5N1, Hemagglutinin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide corresponding to 14aa in the middle region of the Hemagglutinin protein.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Influenza A, Avian, H5N1, Hemagglutinin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: