Anti-CAP1 Rabbit Polyclonal Antibody
100 µl
BIRBORB41759-100
Marketsource
: Biorbyt
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500
Type: Primary
Antigen: CAP1 (CAP, adenylate cyclase-associated protein 1 (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
- Pistosyvyys:100 µl
- Western blot:Yes
- Klonaalisuus:Polyclonal
- Reaktiivisuus:Human
- Säilytyspuskuri:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Kuljetuslämpötila:Ice
- Isäntä:Rabbit
- Pitoisuus:0,92 mg/ml
- ImmunoKemia:Yes
- Immunogeeni:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 266 and 462 of CAP1.
- Cat. No.:BIRBORB41759-100
- Aminohappoluku:266 to 462
- Puhdistus:purified by antigen affinity chromatography
- Vasta-aineen tyyppi:Ensisijainen
- Konjugaatio:Unconjugated
- Antigeenin symboli:CAP1
- Säilytyslämpötila:Cool
- Antigeenin nimi:CAP, adenylate cyclase-associated Protein 1 (yeast)



