Anti-CD41 gpIIb/IIIa Mouse Monoclonal Antibody (Biotin)
Catalog # BIRBORB26370-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:CD41 gpIIb/IIIa
- Clonality:Monoclonal
- Conjugation:Biotin
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:0,01M PBS, pH 7,2, containing 2mM EDTA, 1% BSA
- Storage temperature:Cool
- Concentration:0,1 mg/ml (OD280)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 μg
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD41 gpIIb/IIIa
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human