Anti-Canine Distemper Virus Mouse Monoclonal Antibody
Catalog # BIRBORB24092-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Canine Distemper Virus
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:3,3 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Purified Canine Distemper Virus Envelope Protein
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Canine Distemper Virus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: