Anti-PCNA Mouse Monoclonal Antibody
: Biorbyt
Type: Primary
Antigen: PCNA (proliferating cell nuclear antigen)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa (Mouse)
Reactivity: Human
- Catalog No:
- BIRBORB22894-20
- BIRBORB22894-5
- BIRBORB22894-100
- Antigen symbol:
- PCNA
- PCNA
- PCNA
- Antigen name:
- Proliferating cell nuclear antigen
- Proliferating cell nuclear antigen
- Proliferating cell nuclear antigen
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG3 kappa
- IgG3 kappa
- IgG3 kappa
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Anti-human PCNA mAb is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human PCNA amino acids 1-261 purified from E.coli
- Anti-human PCNA mAb is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human PCNA amino acids 1-261 purified from E.coli
- Anti-human PCNA mAb is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with recombinant human PCNA amino acids 1-261 purified from E.coli
- Amino acid number:
- 1 to 261
- 1 to 261
- 1 to 261
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- purified by Protein G chromatography
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice