Anti-ROBO3 Goat Polyclonal Antibody
Catalog # BIRBORB19560-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Roundabout Guidance Receptor 3
- Antigen symbol:ROBO3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:75 - 80 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:16000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: ROBO3 (roundabout, axon guidance receptor, homolog 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat