Anti-RAX Rabbit Polyclonal Antibody
ABNOPAB0149
: Abnova
- Antibody type:Primary
- Antigen name:Retina and anterior neural fold homeobox
- Antigen symbol:RAX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Gene ID:30062
- Antigen synonyms:Retina and anterior neural fold homeobox protein|Retinal homeobox protein Rx
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: