Order Entry
Finland
ContactUsLinkComponent
Anti-SFXN2 Mouse Polyclonal Antibody
Anti-SFXN2 Mouse Polyclonal Antibody
  ABNOH00118980-B02
 :  Abnova
undefined
Anti-SFXN2 Mouse Polyclonal Antibody
  ABNOH00118980-B02
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Sideroflexin 2
  • Antigen symbol:
    SFXN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    118980
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: SFXN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human