Anti-TP53RK Mouse Monoclonal Antibody [clone: 4B9-1H3]
Catalog # ABNOH00112858-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TP53 regulating kinase
- Antigen symbol:TP53RK
- Clonality:Monoclonal
- Clone:4B9-1H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:112858
- Antigen synonyms:PRPK|C20orf64|BUD32|dJ101A2|Nori-2|Nori-2p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TP53RK
Clonality: Monoclonal
Clone: 4B9-1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human