Anti-IL17F Mouse Monoclonal Antibody [clone: 1E5]
ABNOH00112744-M01
: Abnova
- Antibody type:Primary
- Antigen name:Interleukin F
- Antigen symbol:IL17F
- Clonality:Monoclonal
- Clone:1E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:112744
- Antigen synonyms:cytokine ML-1|mutant IL-17F|IL-17F|CANDF6|interleukin-17F|ML-1|interleukin-24|IL-24|ML1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human