Anti-FOXQ1 Mouse Monoclonal Antibody [clone: 2H6]
ABNOH00094234-M01
: Abnova
- Antibody type:Primary
- Antigen name:Forkhead Box Q1
- Antigen symbol:FOXQ1
- Clonality:Monoclonal
- Clone:2H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:94234
- Antigen synonyms:HFH1|FKHRL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: FOXQ1
Clonality: Monoclonal
Clone: 2H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human