Anti-ZNF461 Mouse Polyclonal Antibody
ABNOH00092283-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 461
- Antigen symbol:ZNF461
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:92283
- Antigen synonyms:HZF28|GIOT-1|GIOT1
- Storage buffer:In 1X PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:ZNF461 (NP_694989.2, 1 a.a. ~ 563 a.a) full-length human protein
- Size:50 µg
- Pk:50 µg
Mouse polyclonal antibody raised against a full-length human ZNF461 protein.
Recommended Dilutons: Western Blot (1 µg/ml).
Type: Primary
Antigen: ZNF461
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human