Anti-NEUROG2 Mouse Monoclonal Antibody [clone: 2A8]
ABNOH00063973-M10
: Abnova
- Antibody type:Primary
- Antigen name:Neurogenin 2
- Antigen symbol:NEUROG2
- Clonality:Monoclonal
- Clone:2A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:63973
- Antigen synonyms:ngn-2|Atoh4|NGN2|bHLHa8|Math4A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NEUROG2
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat