Anti-MS4A12 Mouse Monoclonal Antibody [clone: 4F4]
Catalog # ABNOH00054860-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Membrane-Spanning 4-Domains, Subfamily A, Member 12
- Antigen symbol:MS4A12
- Clonality:Monoclonal
- Clone:4F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:54860
- Antigen synonyms:Ms4a10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MS4A12
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human