Anti-VCX3A Mouse Monoclonal Antibody [clone: 6A3]
ABNOH00051481-M01
: Abnova
- Antibody type:Primary
- Antigen name:Variable charge, X-linked 3A
- Antigen symbol:VCX3A
- Clonality:Monoclonal
- Clone:6A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51481
- Antigen synonyms:VCX8R|VCX-A|VCX3|VCX-8r|VCXA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: VCX3A
Clonality: Monoclonal
Clone: 6A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human