Anti-TFCP2L1 Mouse Polyclonal Antibody
Catalog # ABNOH00029842-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor CP2-like 1
- Antigen symbol:TFCP2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:29842
- Antigen synonyms:CRTR-1|CRTR1|Transcription factor CP2-like protein 1|CP2-related transcriptional repressor 1|TFCP2L1|LBP9|LBP-9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: TFCP2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human