Anti-BLOC1S6 Mouse Monoclonal Antibody [clone: 2H8]
Catalog # ABNOH00026258-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:biogenesis of lysosomal organelles complex-1, subunit 6, pallidin
- Antigen symbol:BLOC1S6
- Clonality:Monoclonal
- Clone:2H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:26258
- Antigen synonyms:PA|PLDN|PALLID|HPS9|BLOS6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BLOC1S6
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human