Anti-GIMAP2 Mouse Monoclonal Antibody [clone: 1E10]
Catalog # ABNOH00026157-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:GTPase IMAP Family member 2
- Antigen symbol:GIMAP2
- Clonality:Monoclonal
- Clone:1E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:26157
- Antigen synonyms:GIMA2_HUMAN.|Immunity associated protein 2|IMAP family member 2|GTPase|HIMAP2|IMAP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GIMAP2
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human