Order Entry
Finland
ContactUsLinkComponent
Anti-PASK Mouse Monoclonal Antibody [clone: 6D10]
Anti-PASK Mouse Monoclonal Antibody [clone: 6D10]
  ABNOH00023178-M01
 :  Abnova
undefined
Anti-PASK Mouse Monoclonal Antibody [clone: 6D10]
  ABNOH00023178-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    PAS Domain Containing serine/threonine kinase
  • Antigen symbol:
    PASK
  • Clonality:
    Monoclonal
  • Clone:
    6D10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    23178
  • Antigen synonyms:
    PAS-Kinase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: PASK
Clonality: Monoclonal
Clone: 6D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human