Anti-MAST2 Mouse Monoclonal Antibody [clone: 3F9]
ABNOH00023139-M01
: Abnova
- Antibody type:Primary
- Antigen name:Microtubule Associated Serine/Threonine Kinase 2
- Antigen symbol:MAST2
- Clonality:Monoclonal
- Clone:3F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23139
- Antigen synonyms:KIAA0807|MTSSK|MAST205
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MAST2
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human