Anti-BAHD1 Mouse Polyclonal Antibody
ABNOH00022893-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Bromo Adjacent Homology Domain Containing 1
- Antigen symbol:BAHD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:22893
- Antigen synonyms:BAH Domain-Containing Protein 1|KIAA0945|Gm117|AL022997|AW541238
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: BAHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human