Order Entry
Finland
ContactUsLinkComponent
 
Anti-SCN11A Mouse Monoclonal Antibody [clone: 2B10]
Catalog # ABNOH00011280-M02
Supplier:  Abnova
undefined
Anti-SCN11A Mouse Monoclonal Antibody [clone: 2B10]
Catalog # ABNOH00011280-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sodium Channel, Voltage-gated, Type XI, Alpha
  • Antigen symbol:
    SCN11A
  • Clonality:
    Monoclonal
  • Clone:
    2B10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    11280
  • Antigen synonyms:
    FEPS3|PN5|NaN|SCN12A|SNS-2|HSAN7|NAV1.9
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SCN11A
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human