Anti-PDIA5 Mouse Monoclonal Antibody [clone: 3A3]
Catalog # ABNOH00010954-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Disulfide Isomerase Family A, Member 5
- Antigen symbol:PDIA5
- Clonality:Monoclonal
- Clone:3A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10954
- Antigen synonyms:PDIR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PDIA5
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human