Anti-TAB1 Mouse Monoclonal Antibody [clone: 2A12]
Catalog # ABNOH00010454-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TGF-beta activated kinase 1
- Antigen symbol:TAB1
- Clonality:Monoclonal
- Clone:2A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10454
- Antigen synonyms:TAB1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|MAP3K7IP1|TAK1-binding protein 1|TGF-beta-activated kinase 1-binding protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TAB1
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat