Anti-BCAS2 Mouse Monoclonal Antibody [clone: 1D10]
ABNOH00010286-M01
: Abnova
- Antibody type:Primary
- Antigen name:Breast carcinoma-amplified sequence 2
- Antigen symbol:BCAS2
- Clonality:Monoclonal
- Clone:1D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:10286
- Antigen synonyms:Breast carcinoma-amplified sequence 2|BCAS2|Pre-mRNA-splicing factor SPF27|SPF27|DAM1|Snt309
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: BCAS2
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human