Anti-TOPORS Mouse Monoclonal Antibody [clone: 6D2]
Catalog # ABNOH00010210-M15A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Topoisomerase I binding, arginine/serine-rich, E3 ubiquitin Protein ligase
- Antigen symbol:TOPORS
- Clonality:Monoclonal
- Clone:6D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10210
- Antigen synonyms:P53BP3|RP31|TP53BPL|LUN
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: TOPORS
Clonality: Monoclonal
Clone: 6D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human