Order Entry
Finland
ContactUsLinkComponent
Anti-HMGXB4 Mouse Monoclonal Antibody [clone: 3B4]
Anti-HMGXB4 Mouse Monoclonal Antibody [clone: 3B4]
  ABNOH00010042-M01
 :  Abnova
undefined
Anti-HMGXB4 Mouse Monoclonal Antibody [clone: 3B4]
  ABNOH00010042-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    HMG Box Domain Containing 4
  • Antigen symbol:
    HMGXB4
  • Clonality:
    Monoclonal
  • Clone:
    3B4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10042
  • Antigen synonyms:
    HMG2L1|THC211630|HMGBCG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: HMGXB4
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human