Anti-CDYL Mouse Monoclonal Antibody [clone: 1A6]
ABNOH00009425-M02
: Abnova
- Antibody type:Primary
- Antigen name:Chromodomain protein, Y-like
- Antigen symbol:CDYL
- Clonality:Monoclonal
- Clone:1A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9425
- Antigen synonyms:CDYL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CDYL
Clonality: Monoclonal
Clone: 1A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human