Anti-MTA1 Mouse Monoclonal Antibody [clone: 1C3]
ABNOH00009112-M10A
: Abnova
- Antibody type:Primary
- Antigen name:Metastasis associated 1
- Antigen symbol:MTA1
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9112
- Antigen synonyms:MTA1|MTA-1|Metastasis Associated Protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: MTA1
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human