Anti-EIF3F Mouse Polyclonal Antibody
ABNOH00008665-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 3, subunit F
- Antigen symbol:EIF3F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8665
- Antigen synonyms:EIF3S5|eIF3-p47
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: EIF3F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human