Anti-SCNN1G Rabbit Polyclonal Antibody
Catalog # ABNOH00006340-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sodium Channel Nonvoltage-gated 1 gamma
- Antigen symbol:SCNN1G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6340
- Antigen synonyms:Epithelial Na(+) channel subunit gamma|ENaCg|SCNEG)|PHA1|Amiloride-sensitive sodium channel subunit gamma (SCNN1G|Gamma-NaCH|SCNEG,|Gamma-ENaC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human