Order Entry
Finland
ContactUsLinkComponent
Anti-S100 A10 Mouse Monoclonal Antibody [clone: 3E10]
Anti-S100 A10 Mouse Monoclonal Antibody [clone: 3E10]
Catalog # ABNOH00006281-M09
Supplier:  Abnova
undefined
Anti-S100 A10 Mouse Monoclonal Antibody [clone: 3E10]
Catalog # ABNOH00006281-M09
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 Calcium Binding Protein A10
  • Antigen symbol:
    S100A10
  • Clonality:
    Monoclonal
  • Clone:
    3E10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6281
  • Antigen synonyms:
    42C|GP11|CLP11|ANX2L|CAL1L|Ca[1]|p10|P11|ANX2LG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: S100 A10
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human