Anti-RPA1 Mouse Polyclonal Antibody
Catalog # ABNOH00006117-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Replication Protein A1
- Antigen symbol:RPA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6117
- Antigen synonyms:HSSB|RF-A|RP-A|MST075|RPA70|REPA1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human