Anti-RFC2 Mouse Polyclonal Antibody
Catalog # ABNOH00005982-B02P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Replication factor C subunit 2
- Antigen symbol:RFC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5982
- Antigen synonyms:Replication factor C 40 kDa subunit|Activator 1 subunit 2|RFC40|replication factor C subunit 2|Activator 1 40 kDa subunit
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: RFC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human