Anti-RARRES2 Mouse Polyclonal Antibody
Catalog # ABNOH00005919-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Retinoic acid receptor responder (tazarotene induced) 2
- Antigen symbol:RARRES2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5919
- Antigen synonyms:TIG2|Chemerin|HP10433|AI303416
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: RARRES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human