Order Entry
Finland
ContactUsLinkComponent
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2B2]
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2B2]
Catalog # ABNOH00005603-M05
Supplier:  Abnova
undefined
Anti-MAPK13 Mouse Monoclonal Antibody [clone: 2B2]
Catalog # ABNOH00005603-M05
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 13
  • Antigen symbol:
    MAPK13
  • Clonality:
    Monoclonal
  • Clone:
    2B2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5603
  • Antigen synonyms:
    MAPK-13|MAPK 13|p38delta|PRKM13|SAPK4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MAPK13
Clonality: Monoclonal
Clone: 2B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human