Anti-SERPINI1 Mouse Monoclonal Antibody [clone: 1E10]
Catalog # ABNOH00005274-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade I, member 1
- Antigen symbol:SERPINI1
- Clonality:Monoclonal
- Clone:1E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5274
- Antigen synonyms:PI12|serpin peptidase inhibitor|SERPINI1|serine (or cysteine) proteinase inhibitor|member 1|clade I (neuroserpin)|neuroserpin|protease inhibitor 12 (neuroserpin)|DKFZp781N13156
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SERPINI1
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human