Anti-SMAD7 Mouse Monoclonal Antibody [clone: 4G11]
ABNOH00004092-M07
: Abnova
- Antibody type:Primary
- Antigen name:SMAD family member 7
- Antigen symbol:SMAD7
- Clonality:Monoclonal
- Clone:4G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4092
- Antigen synonyms:MADH7|SMAD7|Smad7|MADH8|SMAD family member 7|hSMAD7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SMAD7
Clonality: Monoclonal
Clone: 4G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human