Anti-IFIT2 Mouse Polyclonal Antibody
Catalog # ABNOH00003433-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interferon-induced protein with tetratricopeptide repeats 2
- Antigen symbol:IFIT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3433
- Antigen synonyms:cig42|IFIT-2|GARG-39|IFI-54K|G10P2|ISG-54 K|P54|IFI54|IFI-54|ISG-54K|ISG54
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: IFIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human