Anti-HNRNPD Mouse Polyclonal Antibody
Catalog # ABNOH00003184-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein 1, 37kDa)
- Antigen symbol:HNRNPD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3184
- Antigen synonyms:hnRNPD0|AUF1A|HNRPD|AUF1|P37
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: HNRNPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human