Anti-FOXI1 Mouse Polyclonal Antibody
Catalog # ABNOH00002299-B02P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box I1
- Antigen symbol:FOXI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2299
- Antigen synonyms:FKHL10|HFH-3|FREAC6|HFH3|FKH10|FREAC-6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: FOXI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human