Anti-FGR Mouse Monoclonal Antibody [clone: 4C12]
ABNOH00002268-M07
: Abnova
- Antibody type:Primary
- Antigen name:FGR proto-oncogene, Src family tyrosine kinase
- Antigen symbol:FGR
- Clonality:Monoclonal
- Clone:4C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2268
- Antigen synonyms:p55c-fgr|c-fgr|SRC2|p58-Fgr|p55-Fgr|p58c-fgr|c-src2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: FGR
Clonality: Monoclonal
Clone: 4C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human