Anti-CHRNE Mouse Monoclonal Antibody [clone: 6A2]
Catalog # ABNOH00001145-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, nicotinic, epsilon (muscle)
- Antigen symbol:CHRNE
- Clonality:Monoclonal
- Clone:6A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1145
- Antigen synonyms:CMS2A|CMS1D|CMS1E|SCCMS|CMS4C|ACHRE|CMS4A|CMS4B|FCCMS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CHRNE
Clonality: Monoclonal
Clone: 6A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human