Anti-RCBTB2 Mouse Monoclonal Antibody [clone: 4G12]
ABNOH00001102-M06
: Abnova
- Antibody type:Primary
- Antigen name:RCC1 and BTB domain-containing protein 2
- Antigen symbol:RCBTB2
- Clonality:Monoclonal
- Clone:4G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:1102
- Antigen synonyms:CHC1L|RLG
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: RCBTB2
Clonality: Monoclonal
Clone: 4G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human