Anti-XIAP Mouse Monoclonal Antibody [clone: 2B3]
ABNOH00000331-M01
: Abnova
- Antibody type:Primary
- Antigen name:X-linked inhibitor of apoptosis
- Antigen symbol:XIAP
- Clonality:Monoclonal
- Clone:2B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:331
- Antigen synonyms:IAP-like protein|HILP|API3|Baculoviral IAP repeat-containing protein 4|X-linked IAP|X-linked inhibitor of apoptosis protein|XIAP|IAP3|Inhibitor of apoptosis protein 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: XIAP
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human